Spitzkolben, mit Normschliff
Lieferant: Lenz Laborglas GmbH & CO.KG
03222128 03221122 03221128 03221137 03221149 03222122 03222137 03222149
LENZ03222128EA 80.1 CHF
LENZ03222128 271-1232 271-1233 271-1234 LENZ03221149 LENZ03222122 LENZ03222137 LENZ03222149
Spitzkolben, mit Normschliff
Kolben Spitzkolben
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
Order Now
TECHNISCHE DATEN
Environmentally Preferable