6-Aminonicotinonitril 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile

L19282.06 L19282.14
L19282.06EA 63.9 CHF
L19282.06 L19282.14
6-Aminonicotinonitril 98%
6-Aminonicotinonitril

Purity: 98% CAS No.: 4214-73-7 Molecular Formula: C6H5N3 Formula Weight: 119.13 Melting Point: 159-164° Boiling Point: 240-250°/15mm UN No.: 3439 MDL No.: MFCD00128902

Formel: C₆H₅N₃
MW: 119.13 g/mol
Siedepunkt: 240…250 °C (15 mmHg)
Melting Pt: 159…164 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00128902
CAS-Nummer: 4214-73-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR