Trimorpholinophosphine oxide 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Phosphoric Trimorpholide

L16315.06 L16315.14
L16315.06EA 69.6 CHF
L16315.06 L16315.14
Trimorpholinophosphine oxide 99%
Trimorpholinophosphine oxide

Purity: 99% CAS No.: 4441-12-7 Molecular Formula: C12H24N3O4P Formula Weight: 305.32 Melting Point: 188-192° Beilstein Reference: 280155 MDL No.: MFCD00047406

Formel: C₁₂H₂₄N₃O₄P
Melting Pt: 188…192 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00047406
CAS-Nummer: 4441-12-7
EINECS: 224-662-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR