N-Octyl-β-D-glucopyranosid

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

L13259.03 L13259.MD
L13259.03EA 171 CHF
L13259.03 L13259.MD
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid

CAS No.: 29836-26-8 Molecular Formula: C14H28O6 Formula Weight: 292.37 Melting Point: 103-104° Beilstein Reference: 84118 Optical Rotation: -31° (c=2 in methanol)

Formel: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 103…104 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00063288
CAS-Nummer: 29836-26-8
EINECS: 249-887-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR