2-Bromnonan ≥97%

Lieferant: Thermo Fisher Scientific

L07387.03 L07387.06
L07387.03EA 61 CHF
L07387.03 L07387.06
2-Bromnonan ≥97%
2-Bromnonan

Purity: 97% CAS No.: 2216-35-5 Molecular Formula: C9H19Br Formula Weight: 207.16 Refractive Index: 1.4530 Boiling Point: 208-209° MDL No.: MFCD00017865

Formel: H₃CBrCH(CH₂)₆CH₃
MW: 207.15 g/mol
Siedepunkt: 208…209 °C
Dichte: 1.081
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00017865
CAS-Nummer: 2216-35-5
EINECS: 218-688-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR