4,4''-Diamino-p-terphenyl 95%

Lieferant: Thermo Fisher Scientific
Achtung

L05403.03 L05403.06
L05403.03EA 167 CHF
L05403.03 L05403.06
4,4''-Diamino-p-terphenyl 95%
4,4''-Diamino-p-terphenyl

Purity: 95% CAS No.: 3365-85-3 Molecular Formula: C18H16N2 Formula Weight: 260.34 Melting Point: 244-250° Beilstein Reference: 2120295

Formel: C₁₈H₁₆N₂
MW: 260.34 g/mol
Melting Pt: 244…250 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00051532
CAS-Nummer: 3365-85-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR