5-Amino-2,4(1H,3H)-pyrimidindion 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Aminouracil, 5-Aminopyrimidine-2,4(1H,3H)-dione

L04452.06 L04452.14
L04452.06EA 54.9 CHF
L04452.06 L04452.14
5-Amino-2,4(1H,3H)-pyrimidindion 97%
5-Aminouracil

Purity: 97% CAS No.: 932-52-5 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Melting Point: 300° Beilstein Reference: 127250

Formel: C₄H₅N₃O₂
MW: 127.1 g/mol
Siedepunkt: 377
Melting Pt: >300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006025
CAS-Nummer: 932-52-5
EINECS: 213-252-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR