2-Brom-1,3-diethylbenzol ≥94%

Lieferant: Thermo Fisher Scientific
Achtung

L03482.03 L03482.06
L03482.03EA 91.3 CHF
L03482.03 L03482.06
2-Brom-1,3-diethylbenzol ≥94%
2-Brom-1,3-diethylbenzol

Purity: 94% CAS No.: 65232-57-7 Molecular Formula: C10H13Br Formula Weight: 213.12 Refractive Index: 1.5440 Boiling Point: 78-80°/2mm MDL No.: MFCD00017770

Formel: C₁₀H₁₃Br
Siedepunkt: 78…80 °C (2 mmHg)
Dichte: 1.264
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00017770
CAS-Nummer: 65232-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR