Cholesteryl-pelargonat

Lieferant: Thermo Fisher Scientific

Synonyms: Cholesterol Nonanoate, Nonanoic Acid Cholesterol Ester, Pelargonic Acid Cholesterol Ester

L02857.14 L02857.22
L02857.14EA 67.1 CHF
L02857.14 L02857.22
Cholesteryl-pelargonat
Cholesterol pelargonate

CAS No.: 1182-66-7 Molecular Formula: C36H62O2 Formula Weight: 526.89 Odour: Odourless Melting Point: 75-79° Beilstein Reference: 3179820 Optical Rotation: -30° (c=5 in chloroform) MDL No.: MFCD00003643

Formel: C₃₆H₆₂O₂
MW: 526.89 g/mol
Melting Pt: 75…79 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003643
CAS-Nummer: 1182-66-7
EINECS: 214-658-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR