Fire extinguisher signs
Lieferant: KROSCHKE
F18323
KROSF18323EA 1360 CHF
KROSF18323
Fire extinguisher signs
Warn- und Verbotszeichen
These are fire extinguisher signs with holes and holders.
  • Temperature resistant from −30 to +50 °C
  • Resistant to moisture and weak acids