Bulb pipettes, with safety bulb, Qualicolor, colour-coded, one mark, Class AS
Lieferant: Kavalier
632433311025
KAVA632433311025EA 32.6 CHF
KAVA632433311025
Bulb pipettes, with safety bulb, Qualicolor, colour-coded, one mark, Class AS
Pipetten Vollpipetten
Soda-lime glass.
  • Enlarged form
  • Not autoclavable at 121 °C
Zertifizierungen: DIN 12691, ISO 648