Filtering flasks with glass or plastic hose connection
Lieferant: Kavalier
632412021950_U 632412022500 632412022956 632412022966 632412021100 632412021250 632412021500 632412022100 632412022250_U 632412021940 632412021956 632412022950 412022940 412022500 632412022940
KAVA632412021950_UEA 128 CHF
KAVA632412021950_U KAVA632412022500 KAVA632412022956 KAVA632412022966 KAVA632412021100 KAVA632412021250 KAVA632412021500 KAVA632412022100 KAVA632412022250_U KAVA632412021940 KAVA632412021956 KAVA632412022950 KAVA412022940 KAVA412022500 KAVA632412022940
Filtering flasks with glass or plastic hose connection
Kolben Saugflaschen
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flask filtering with plastic or glass side hose connection.