Notrax® 562 Sanitop® Floor Mattings
Lieferant: JUSTRITE MANUFACTURING
562S0035BL
JUSU562S0035BLEA 62.6 CHF
JUSU562S0035BL
Notrax® 562 Sanitop® Floor Mattings
Fußmatten Anti-Ermüdungsmatten
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
  • Durable rubber compound
  • Molded alternating large/small hole drainage system keeps moisture and debris free from work area
  • Raised pattern surrounds large holes to channel drainage and provide worker traction
  • Beveled ramps on all 4 sides
  • 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards.More Product Information