Horizontal fold flat disposable masks, Springfit™
Lieferant: JSP
BGA122-202-000 BGA172-202-000 BGA132-206-000 BGA182-206-000 BGA802-206-000
JSPABGA122-202-000EA 46.3 CHF
JSPABGA122-202-000 JSPABGA172-202-000 JSPABGA132-206-000 JSPABGA182-206-000 JSPABGA802-206-000
Horizontal fold flat disposable masks, Springfit™
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
The SpringFit™ fold flat disposable mask achieves an unrivalled secure fit with the use of a unique endoskeleton structure. SpringFit™ introduces an abundance of features that maximise fit, comfort, compatibility, and performance.
  • Fold flat disposable mask
  • Compact and easily pocketed
  • Unique endoskeleton structure
  • Polypropylene inner skeleton
  • Available in FFP3 and FFP2, valved and unvalved
The SpringFit™ fold flat disposable mask is available in FFP3 and FFP2, valved and unvalved, with a nuisance odour carbon layer available on the FFP3 model. TheMore Product Information