Silber 0,02 mg/ml sodium citrate_2mM_supplied, nanoparticles 80 nm (Abs. 457 nm)

Lieferant: Thermo Fisher Scientific

J67252.ACEA 176 CHF
J67252.AC J67252.AE
Silber 0,02 mg/ml sodium citrate_2mM_supplied, nanoparticles 80 nm (Abs. 457 nm)
Silver

CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397

Formel: Ag
MW: 107.87 g/mol
Siedepunkt: 2212
Melting Pt: 962
Lager Temperatur: Kühlschrank
MDL Number: MFCD00003397
CAS-Nummer: 7440-22-4
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR