Dinatriumphosphonomycin 90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Disodium (1R,2S)-1,2-Epoxypropylphosphonate, Phosphomycin Disodium Salt, (1R,2S)-1,2-Epoxypropylphosphonic Acid Disodium Salt

J66602.14
J66602.14EA 767 CHF
J66602.14
Dinatriumphosphonomycin 90%
Fosfomycin disodium salt

Prevents peptidoglycan synthesis by inhibiting MurA, an enzyme responsible for synthesizing N-acetylmuramic acid or NAM, a major component of peptidoglycan


  • White to off-white powder


Freely soluble in water or aqueous solution

Formel: C₃H₅Na₂O₄P
MW: 182.02 g/mol
Melting Pt: >300 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00056853
CAS-Nummer: 26016-99-9
EINECS: 247-409-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR