PJ-34 hydrochloride

Lieferant: Thermo Scientific

Synonyms: 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride, PJ34 HCl

J64413.MB
J64413.MBEA 1210 CHF
J64413.MB
PJ-34 hydrochloride
PJ-34 hydrochloride

Applications: A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase

Formel: C₁₇H₁₇N₃O₂·HCl
MW: 295,2·36,5 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD22581510
CAS-Nummer: 344458-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR