3'-Amino-2',3'-dideoxythymidine 99%

Lieferant: Thermo Fisher Scientific

J64342.03
J64342.03EA 99.7 CHF
J64342.03
3'-Amino-2',3'-dideoxythymidine 99%
3'-Amino-2',3'-dideoxythymidine

Purity: 99% CAS No.: 52450-18-7 Molecular Formula: C10H15N3O4 Formula Weight: 241.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Off-white to yellow MDL No.: MFCD00038057

Formel: C₁₀H₁₅N₃O₄
MW: 241.24 g/mol
MDL Number: MFCD00038057
CAS-Nummer: 52450-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR