Doxorubicin Hydrochlorid

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

J64000.MA J64000.MF
J64000.MAEA 399 CHF
J64000.MA J64000.MF
Doxorubicin Hydrochlorid
Doxorubicin hydrochloride

Doxorubicin hydrochloride is an anti-tumour, anti-neoplastic and anti-biotic agent that inhibits DNA topoisomerase II. It also inhibits nucleic acid synthesis and induces apoptosis.


  • Solid form
  • Soluble in water


Doxorubicin hydrochloride possesses fluorescent properties, which enable it to be used for the measurement of drug efflux pump activities and investigating the role of subcellular organelles (golgi and lysosome) in the sequestration of drugs.


Bitte beachten Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidising agents.

Formel: C₂₇H₂₉NO₁₁·HCl
MW: 579.98 g/mol
Melting Pt: 216 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00077757
CAS-Nummer: 25316-40-9
EINECS: 246-818-3
Merck Index: 14,3439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR