Carbonate-buffered saline (5X), pH 9,5

Lieferant: Thermo Fisher Scientific

J63928.AP
J63928.APEA 110 CHF
J63928.AP
Carbonate-buffered saline (5X), pH 9,5
Puffer Gewebekultur-Puffer

CAS No.: 497-19-8
Physical Form: Liquid
MDL No.: MFCD00003494

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR