Ethacrynic acid

Lieferant: Thermo Fisher Scientific
Achtung

J63684.03
J63684.03EA 164 CHF
J63684.03
Ethacrynic acid
Ethacrynic acid

CAS No.: 58-54-8 Molecular Formula: C13H12Cl2O4 Formula Weight: 303.14 Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00056693


Applications: Diuretic that acts by inhibiting sodium-potassium-chloride cotransport

Formel: C₁₃H₁₂Cl₂O₄
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00056693
CAS-Nummer: 58-54-8
EINECS: 200-384-1
Merck Index: 14,3717

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR