Sodium pyrophosphate, 200 mM buffer solution

Lieferant: Thermo Fisher Scientific

J62052.AE J62052.AK
J62052.AEEA 131 CHF
J62052.AE J62052.AK
Sodium pyrophosphate, 200 mM buffer solution
Puffer Klonpuffer

CAS No.: 7758-16-9
Physical Form: Liquid
MDL No.: MFCD00014246

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR