Buffer, sodium acetate, 3 M aq, soln, pH 5,2, RNAse free
Lieferant: Thermo Fisher Scientific
J61928.AK J61928.AE
J61928.AKEA 220 CHF
J61928.AK J61928.AE
Buffer, sodium acetate, 3 M aq, soln, pH 5,2, RNAse free
Puffer Klonpuffer
Sodium acetate buffer solution pH 5,2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis.
  • RNase-free
Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.