IEF Cathode buffer (10X) with lysine

Lieferant: Thermo Fisher Scientific

J61903.AK
J61903.AKEA 74.2 CHF
J61903.AK
IEF Cathode buffer (10X) with lysine
Puffer Elektrophorese-Puffer Laufpuffer

Physical Form: Liquid
Appearance: Colourless
Applications: For isoelectric focusing (IEF)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR