Uranin

Lieferant: Thermo Fisher Scientific

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

J61549.22 J61549.36
J61549.22EA 70 CHF
J61549.22 J61549.36
Uranin
Uranin

CAS No.: 518-47-8 Molecular Formula: C20H10Na2O5 Formula Weight: 376.28 Physical Form: Powder Appearance: Dark brownish-red to dark Solubility: Soluble in water at 1mg/ml Beilstein Reference: 3833041 Merck Reference: 14,4159 MDL No.: MFCD00167039


A fluorophore comonly used in microscopy

Formel: C₂₀H₁₀Na₂O₅
Melting Pt: > 300 °C
Dichte: 1.601 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00167039
CAS-Nummer: 518-47-8
EINECS: 208-253-0
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR