Kanamycinsulfat

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

J61272.09 J61272.14 J61272.22
J61272.09EA 244 CHF
J61272.09 J61272.14 J61272.22
Kanamycinsulfat
Kanamycin sulphate

CAS No.: 25389-94-0 Molecular Formula: C18H36N4O11.H2SO4 Formula Weight: 582.58 Physical Form: Powder Appearance: White to off-white Solubility: Dissolve in water at 10mg/ml MDL No.: MFCD00070253


Antibiotic. Inhibits translocation during protein biosynthesis

Formel: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00070253
CAS-Nummer: 25389-94-0
EINECS: 246-933-9

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR