Theophyllin

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

J60203.5H J60203.RX
J60203.5HEA 150 CHF
J60203.5H J60203.RX
Theophyllin
Theophyllin
Formel: C₇H₈N₄O₂
MW: 180.16 g/mol
Melting Pt: 270…274 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00079619
CAS-Nummer: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1 III
Merck Index: 14,9285

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR