Gravimetric diluters, DiluFlow® Pro
Lieferant: INTERSCIENCE
502105 502205
INSC502105EA 0 CHF
INSC502105 INSC502205
Gravimetric diluters, DiluFlow® Pro
Verdünner Gravimetrische Verdünner
DiluFlow® Pro automatically dilutes in seconds a solid sample with the appropriate weight of diluent with traceability. With its robotic arm and high speed process, it offers an efficient and traceable solution for the dilution process of microbiological analyses.
  • Ultra low profile: bag opening height is only 30 cm which makes it ideal for work in a laminar flow cabinet
  • GeckoGrip System: Holds the bag in all conditions
  • Light Code: Intuitive indicator of the status
  • Removable DripTray for easy cleaning, in case of spillage
  • Distribution and weighing functions
More Product Information