Anti-IgG Donkey Polyclonal Antibody (DyLight® 550)
Lieferant: ImmunoReagents
IR1286
IMMRIR1286EA 303 CHF
IMMRIR1286
Anti-IgG Donkey Polyclonal Antibody (DyLight® 550)
Antikörper
The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multiColour applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562nm/576nm Emission Colour = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3,More Product Information