Anti-PAI-1 (Ab-1) (2) Mouse Monoclonal Antibody [clone: 2]

Lieferant: Merck Millipore (Oncogene)
IM29L-100
IM29L-100EA 1200 CHF
IM29L-100
Anti-PAI-1 (Ab-1) (2) Mouse Monoclonal Antibody [clone: 2]
Antikörper
Type: Primary
Antigen: PAI-1 (Ab-1) (2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR