Oil Free Turbomolecular Pumping Systems, CDK
Lieferant: Welch by Gardner Denver
101251 101250
ILMV101251EA 12260 CHF
ILMV101251 ILMV101250
Oil Free Turbomolecular Pumping Systems, CDK
Pumpen Vakuum-Membranpumpen
CDK turbomolecular pumping systems are completely oil-free. It incorporate an oil-free SST turbomolecular pump using dry-running, solid-lubricated ceramic bearings. The backing pump is an oil-free diaphragm vacuum pump.
  • Unique, built-in automatic shut-off device for the backing pump
  • User friendly
  • Automatic shut-off device for the backing pump
  • Lightweight, compact and portable
If the pump system works continually at the ultimate pressure, and if the connected vacuum system has a low real and virtual leak rate (wall degassing), then it is possible thatMore Product Information