Lysing kits for Fastprep®-24 5G, Lysing Matrix E, MP Biomedicals
Lieferant: MP Biomedicals
116934050 116984010 116914500 116914100 116914050 116934025 116954010 116954050 116974025 116974050 116974100
ICNA116934050EA 652 CHF
ICNA116934050 ICNA116984010 ICNA116914500 ICNA116914100 ICNA116914050 ICNA116934025 ICNA116954010 ICNA116954050 ICNA116974025 ICNA116974050 ICNA116974100
Lysing kits for Fastprep®-24 5G, Lysing Matrix E, MP Biomedicals
Homogenisatoren Tischhomogenisatoren
Each impact-resistant 2 ml tube contains 1,4 mm ceramic spheres, 0,1 mm silica spheres, and one 4 mm glass bead. Lysing Matrix E tubes have purple caps and found in the FastDNA™ SPIN Kit for Soil and the FastRNA™ Pro Soil Kits. Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumours and other difficult tissues.
Order Now
TECHNISCHE DATEN
Environmentally Preferable