Hand-pump for bottles, MP Biomedicals
Lieferant: MP Biomedicals
0976680HP
ICNA0976680HPEA 15.2 CHF
ICNA0976680HP
Hand-pump for bottles, MP Biomedicals
Pumpen Fass/Transferpumpen
White hand pump suitable for 7X bottles.
  • Can be store at room temperature (15 to 30 °C)
  • This pump is DNase, RNase and protease free
  • Can be used in all research areas
Dispenses 29.57 ml per press.
Bitte beachten Research use only (RUO). Ready for CE IVD labeling of clinical applications.