MacConkey agar, MP Biomedicals
Lieferant: ICN Biomedicals
091006017
ICNA091006017EA 147 CHF
ICNA091006017
MacConkey agar, MP Biomedicals
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
A preparation for the isolation of enteric bacilli based on the fermentation of lactose.
Ready for CE IVD labeling of clinical applications.
Bitte beachten Research use only (RUO).