Freund's incomplete adjuvant, MP Biomedicals
Lieferant: ICN Biomedicals
08642861
ICNA08642861EA 175 CHF
ICNA08642861
Freund's incomplete adjuvant, MP Biomedicals
Protein- Expression Proteinexpressions-Reagenzien
Freund's incomplete adjuvant mixture contains mineral oil and Arlacel A.
  • Enhances immune response in booster injections
  • Water-in-oil emulsion without Mycobacterium