D(+)-Saccharose, Kristalle, für die Nahrungsmittelindustrie

Lieferant: ICN Biomedicals

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

0290471305 0290471325 0290471380
ICNA0290471305EA 72.8 CHF
ICNA0290471305 ICNA0290471325 ICNA0290471380
D(+)-Saccharose, Kristalle, für die Nahrungsmittelindustrie
D(+)-Saccharose

Soluble in water (1 g/0,5 ml at 25 °C; 1 g/0,2 ml boiling water), ethanol (1 g/170 ml) or methanol (1 g/100 ml); moderately soluble in glycerol or pyridine.

Formel: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 177…187 °C
MDL Number: MFCD00006626
CAS-Nummer: 57-50-1
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR