HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)

Lieferant: ICN Biomedicals

Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

0219454910 0219454950 0219454983 0219454991 0219454925 0219454980 0219454990
ICNA0219454910EA 49 CHF
ICNA0219454910 ICNA0219454950 ICNA0219454983 ICNA0219454991 ICNA0219454925 ICNA0219454980 ICNA0219454990
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)

Synonym: N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid) Cell Culture Reagent Crystalline Free Acid Zwitterionic Buffer useful in the pH range 6.8-8.2. pKa=7.55 at 25°C


pH: 4.0 - 5.5 (1% aq soln)

Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Melting Pt: 210…215 °C
Dichte: 1.448 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006158
CAS-Nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR