Acriflavin Hydrochlorid, powder_redish brown

Lieferant: ICN Biomedicals
Gefahr

Synonyms: 3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

0219067510 0219067525 0219067580
ICNA0219067510EA 67.5 CHF
ICNA0219067510 ICNA0219067525 ICNA0219067580
Acriflavin Hydrochlorid, powder_redish brown
Acriflavin Hydrochlorid

Acriflavine Hydrochloride is a mixture of 3,6-diamino-10-methylacridinium chloride and 3,6-diaminoacridine (approximately an 9 to 1 mixture respectively).


  • UV/Visible Absorbance: λ max (meOH) 460 ± 4 nm
  • Soluble in water (1 mg/ml- clear, dark orange solution).


Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

Formel: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
Melting Pt: 260 °C (Zers., Lit.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00069039
CAS-Nummer: 8063-24-9
UN: 3077
ADR: 9 III

Order Now

Spezifikation Testergebnisse

Identity Test Passes
UV/Vis: l max (meOH) 460±4nm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR