Dextrin, weißes Pulver Commercial Grade, Type III

Lieferant: ICN Biomedicals

Synonyms: Dextrin

0216005790
ICNA0216005790EA 45.3 CHF
ICNA0216005790
Dextrin, weißes Pulver Commercial Grade, Type III
Dextrin

Dextrin is used in nutrition studies to understand the effects of fibre in diets on energy levels, intestinal flora, digestion and other physiological processes.


  • Calories: Contains approximately 4,12 Kcal/g dry weight (Lit.)


Dextrin is used in nutrition studies to understand the effects of fiber in diets on energy levels, intestinal flora, digestion and other physiological processes.


Prepared from amylopectin by Hydrochloric acid hydrolysis: the amylopectin is roasted in vessels jacketed in hot oil - the oil is heated to 450 °C - at the same time HCl is added to the amylopectin - the acid plus the heat causes the molecular structure of the starch to change/polymerise. As the hydrolysis progresses, water solubility increases and color might develop.


Bitte beachten Research Use Only (RUO).

Formel: (C₆H₁₂O₆)n
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081554
CAS-Nummer: 9004-53-9

Order Now

Spezifikation Testergebnisse

Identity Test Passes
Source Corn
Moisture Content 13% maximum
pH (12% aq. soln.) 3.5 - 4.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR