Sesame oil, farbloses Öl

Lieferant: ICN Biomedicals

0215662183 0215662191
ICNA0215662183EA 107 CHF
ICNA0215662183 ICNA0215662191
Sesame oil, farbloses Öl
Sesame oil

Sesame oil is a potent antioxidant, which can inhibit DNA damage and lipid peroxidation by 4-Nitroquinoline N-oxide in rats.


Studies indicate that Sesame oil can increase wound healing in SKH1 mice when added together with ozone. It is known that Sesame oil is composed of triacylglycerols encapsulated by a monolayer of phospholipids which is then covered by the proteins oleosin, caleosin and steroleosin. Furthermore, this compound is a great research tool for reversing and preventing oxidative damage on cells and tissues.
Sesame oil used in research applications as delivery vehicle for fat-soluble compounds including steroidal hormones and toxins. Sesame oil has antioxidant properties and was shown to protect against lipopolysaccharide (LPS)-stimulated oxidative stress in animal models.


Soluble in chloroform, ether, petroleum ether, carbon disulphide; slightly soluble in alcohol; insoluble in water.

MDL Number: MFCD00132243
CAS-Nummer: 8008-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR