2-(4-Nitrophenyl)ethanol, Pulver

Lieferant: ICN Biomedicals
Achtung

Synonyms: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol

0215589005 0215589050 0215589010
ICNA0215589005EA 90 CHF
ICNA0215589005 ICNA0215589050 ICNA0215589010
2-(4-Nitrophenyl)ethanol, Pulver
2-(4-Nitrophenyl)ethanol

p-Nitrophenethyl alcohol is used as a protecting reagent for phosphates and O6 of deoxyguanosine.


Soluble in acetone (50 mg/ml) and methanol (25 mg/ml).

Formel: C₈H₉NO₃
Melting Pt: 59…62 °C (lit.)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00010202
CAS-Nummer: 100-27-6
EINECS: 202-835-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR