Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Lieferant: ICN Biomedicals
0210330401
ICNA0210330401EA 94.1 CHF
ICNA0210330401
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Medien für die Mikrobiologie Basismaterialien für die Mikrobiologie
Yeast Hydrolysate Enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components, carbohydrates, and B complex vitamins natural to yeast.
  • Soluble in water
  • Standard plate count: <50,000/g
  • Protein (N×6,25): 50,0 to 65,0%