Pectin, Pulver

Lieferant: ICN Biomedicals

0210258791 0210258780 0210258790
ICNA0210258791EA 0 CHF
ICNA0210258791 ICNA0210258780 ICNA0210258790
Pectin, Pulver
Pectin

Soluble in water (1 g/20 ml - viscous solution acid to litmus); insoluble in organic solvents.


Pectin is a heterosaccharide, component of terrestrial plant cell walls.


It is used as a substrate to identify, differentiate and characterised pectinase(s). It is also used to study its degradation by pectinolytic bacteria.

MDL Number: MFCD00081838
CAS-Nummer: 9000-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR