Indophenol, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: Phenolindophenol

M-Clarity™ quality level = MQ100

I5763-1G I5763-5G
I5763-1GEA 84.7 CHF
I5763-1G I5763-5G
Indophenol, Sigma-Aldrich®
Indophenol

Indophenol is used in hair dyes, redox materials, lubricants, liquid crystal displays, biosensor and fuel cells. It is toxic to fishes and is implicated in environmental pollution. Indophenol method is common for the determination of ammonia. The reaction gives a blue product, which is measured spectrophotometrically.

Formel: C₁₂H₉NO₂
Melting Pt: >300 °C (lit.)
Lager Temperatur: Room Temperature
MDL Number: MFCD00001621
CAS-Nummer: 500-85-6
EINECS: 207-913-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d99604379c6c52c6d0236b0f800257236fea44c4&vid=06990a56bcbe0fe607176c9fdc9e67b7af310abe&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Colour) Dark Green to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour) Blue to Dark Blue to Dark Green
Solubility (Turbidity), 10 mg/ml, 1 M NaOH Clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR