Aflatoxin G2-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Lieferant: Honeywell Chemicals
R1959-2ML
HONCR1959-2MLEA 286.07 CHF
HONCR1959-2ML
Aflatoxin G2-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Standards Organische Standards
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery