Copper AAS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4022-100ML E4022-500ML E4023-100ML E4023-500ML E4109-100ML E4201-100ML E4201-500ML
HONCE4022-100MLEA 40.23 CHF
HONCE4022-100ML HONCE4022-500ML HONCE4023-100ML HONCE4023-500ML HONCE4109-100ML HONCE4201-100ML HONCE4201-500ML
Copper AAS Standards, (CRMs), Fluka™
Standards Kationischer Standards Kupfer-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).