Succinate Standard for Ion Chromatography, (CRM), Fluka™
Lieferant: Honeywell Chemicals
E3046-100ML
HONCE3046-100MLEA 61.52 CHF
HONCE3046-100ML
Succinate Standard for Ion Chromatography, (CRM), Fluka™
Standards Anionische Standards Succinate-Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).