Multi-element standard solution 1 for ICP
Lieferant: Honeywell Chemicals
E2230-100ML
HONCE2230-100MLEA 188.05 CHF
HONCE2230-100ML
Multi-element standard solution 1 for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
100 mg/L each of Bi, K, Pb, 50 mg/L each of Al, B, Cr, Li, Mo, Na, Ni, Tl, 10 mg/L each of Ag, Ba, Ca, Cd, Co, Cu, Fe, Mg, Mn, Sr, Zn in HNO₃ 10%