Instrument calibration standard 1 for ICP
Lieferant: Honeywell Chemicals
E2148-100ML
HONCE2148-100MLEA 448.42 CHF
HONCE2148-100ML
Instrument calibration standard 1 for ICP
Standards Kationischer Standards Mehrelement-Standards
Certified Reference Material (CRM).
20 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn in HNO₃ 5%