Aluminium ICP & ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1301-100ML E1370-100ML
HONCE1301-100MLEA 114.07 CHF
HONCE1301-100ML HONCE1370-100ML
Aluminium ICP & ICP-MS Standards, (CRMs), Fluka™
Standards Kationischer Standards Aluminium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Zertifizierungen: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).